Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Catalog NumberQuantity
ALF088057.AP500 mL
Catalog number ALF088057.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7783-83-7
IUPAC Nameiron(3+) ammonium dodecahydrate disulfate
Molecular FormulaFeH28NO20S2
InChI KeyLCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Potassium (K)=<0.005 %
Chloride (Cl)=<0.001 %
Copper (Cu)=<0.003 %
Appearance (Form)Adhering powder or crystals
Appearance (Color)Violet