Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
For Use in the Manufacture of Fertilizers and Explosives, in organic Synthesis, in Metallurgy
Have Questions?
Catalog NumberQuantity
ALF087920.AP500 mL
Catalog number ALF087920.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNitric Acid
ColorColorless to Yellow
CAS7697-37-2
Boiling Point121°C
Health Hazard 1H272-H290-H314-H331
View more
In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuelsMain commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.