Vanadium(V) trichloride oxide, V+5 28.5% min, Thermo Scientific Chemicals
Vanadium(V) trichloride oxide, V+5 28.5% min, Thermo Scientific Chemicals
Vanadium(V) trichloride oxide, V+5 28.5% min, Thermo Scientific Chemicals
Vanadium(V) trichloride oxide, V+5 28.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vanadium(V) trichloride oxide, V+5 28.5% min, Thermo Scientific Chemicals
CAS: 7727-18-6 | Cl3OV | 173.29 g/mol
Have Questions?
Catalog NumberQuantity
ALF081118.22100 g
Catalog number ALF081118.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS76-89-1
IUPAC Namemethyl 2-hydroxy-2,2-diphenylacetate
Molecular FormulaC15H14O3
InChI KeyLJFIHTFNTGQZJL-UHFFFAOYSA-N
SMILESCOC(=O)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)71.0-77.0?C
Vanadium(V) trichloride oxide is used as a catalyst in olefin polymerization reaction, especially for the production of ethylene-propylene rubber. It is also used to synthesis organovanadium compounds and as a mordant in dyeing. It acts as a precursor to prepare vanadium pentoxide, vanadyl dichloride and dioxovanadium monochloride. Further, it serves as a reagent and a strong oxidizing agent in organic synthesis.