Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals
Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals
Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals
Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals
CAS: 3087-36-3 | C8H20O4Ti | 228.111 g/mol
Have Questions?
Catalog NumberQuantity
ALF077142.1850 g
Catalog number ALF077142.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS128-69-8
IUPAC Name7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular FormulaC24H8O6
InChI KeyCLYVDMAATCIVBF-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<1 % (K.F.)
Assay>=97.5 %
Appearance (Color)Dark red
Infrared spectrumConforms
Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.