Diphenyltin dichloride, 90+%, Thermo Scientific Chemicals
Diphenyltin dichloride, 90+%, Thermo Scientific Chemicals
Diphenyltin dichloride, 90+%, Thermo Scientific Chemicals
Diphenyltin dichloride, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diphenyltin dichloride, 90+%, Thermo Scientific Chemicals
CAS: 1135-99-5 | C12H12Cl2Sn | 345.84 g/mol
Have Questions?
Catalog NumberQuantity
ALF071139.0910 g
Catalog number ALF071139.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.
Used in the manufacture of chemicals.