Tin(IV) iodide, 95%, Thermo Scientific Chemicals
Tin(IV) iodide, 95%, Thermo Scientific Chemicals
Tin(IV) iodide, 95%, Thermo Scientific Chemicals
Tin(IV) iodide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tin(IV) iodide, 95%, Thermo Scientific Chemicals
CAS: 7790-47-8 | H4I4Sn | 630.36 g/mol
Have Questions?
Catalog NumberQuantity
ALF071114.0910 g
Catalog number ALF071114.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS119-30-2
IUPAC Name2-hydroxy-5-iodobenzoic acid
Molecular FormulaC7H5IO3
InChI KeySWDNKOFGNPGRPI-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(I)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Identification (FTIR)Conforms
View more
Used in chemical research and as a food additive.