Dimethyl selenide, Thermo Scientific Chemicals
Dimethyl selenide, Thermo Scientific Chemicals
Dimethyl selenide, Thermo Scientific Chemicals
Dimethyl selenide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl selenide, Thermo Scientific Chemicals
CAS: 593-79-3 | C2H6Se | 109.041 g/mol
Have Questions?
Catalog NumberQuantity
ALF068116.031 g
Catalog number ALF068116.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS4023-34-1
IUPAC Namecyclopropanecarbonyl chloride
Molecular FormulaC4H5ClO
InChI KeyZOOSILUVXHVRJE-UHFFFAOYSA-N
SMILESClC(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Titration RCOCl>=97.5 %
Refractive index1.4515 to 1.4535 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
Used as primary and secondary intermediate and also used in chemical research.