Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
CAS: 3425-46-5 | CKNSe | 144.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF068107.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium selenocyanate
Melting Point162°C to 164°C
CAS3425-46-5
ColorWhite
Health Hazard 1H301+H331-H373
View more
Potassium selenocyanate is used to prepare triphenylphosphine selenide by reacting with triphenylphosphine. It finds application in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury thiocyanate and selenocyanate compounds.