Sodium sulfide, anhydrous, Thermo Scientific Chemicals
Sodium sulfide, anhydrous, Thermo Scientific Chemicals
Sodium sulfide, anhydrous, Thermo Scientific Chemicals
Sodium sulfide, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium sulfide, anhydrous, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF065122.065 g
Catalog number ALF065122.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium sulfide, anhydrous
Melting Point950°C
CAS1313-82-2
Health Hazard 1H290-H301+H311-H314-H500
Health Hazard 3P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
View more
In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagentSodium sulfide is used mainly in the pulp and paper industry in the Kraft process. It serves as an oxygen scavenger agent in water treatment. Other applications include ore flotation, oil recovery, making dyes and detergents. In the leather processing industry, it is utilized for liming operation as an unhairing agent. It is also used in the textile industry as a bleaching and desulfurizing agent and as a dechlorinating agent in chemical photography for toning black and white photographs as well as metals precipitant. It plays an important role as a sulfonation and sulfomethylation agent in the chemical industry and in the production of rubber chemicals, sulfur dyes and other chemical compounds.