Sodium peroxydisulfate, 98%, Thermo Scientific Chemicals
Sodium peroxydisulfate, 98%, Thermo Scientific Chemicals
Sodium peroxydisulfate, 98%, Thermo Scientific Chemicals
Sodium peroxydisulfate, 98%, Thermo Scientific Chemicals
Sodium peroxydisulfate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium peroxydisulfate, 98%, Thermo Scientific Chemicals
CAS: 7775-27-1 | Na2O8S2 | 238.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number ALF054100.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium peroxydisulfate
CAS7775-27-1
Health Hazard 1H272-H302-H315-H317-H319-H334-H335
Health Hazard 3P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
Recommended StorageAmbient temperatures
View more
Sodium peroxydisulfate is used as a radical initiator for emulsion polymerization reactions like acrylonitrile butadiene styrene, detergent component, soil conditioner and soil remediation. It is also used for curing of formaldehyde adhesives. It acts as a bleaching agent and in the production of dyestuffs. It finds application in zinc and printed circuit boards and considered to be a replacement for ammonium persulfate in etching mixtures. Further, it is used in the preparation of diapocynin from apocynin. It plays a role in the conversion of phenols to para-diphenols in alkaline solution and of arylamines to aminophenols. It is actively involved in the Elbs persulfate oxidation and the Boyland-Sims oxidation reactions as an oxidizing agent.