Nickel(II) tetrafluoroborate hexahydrate, Thermo Scientific Chemicals
Nickel(II) tetrafluoroborate hexahydrate, Thermo Scientific Chemicals
Nickel(II) tetrafluoroborate hexahydrate, Thermo Scientific Chemicals
Nickel(II) tetrafluoroborate hexahydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel(II) tetrafluoroborate hexahydrate, Thermo Scientific Chemicals
CAS: 15684-36-3 | B2F8H12NiO6 | 340.39 g/mol
Have Questions?
Catalog NumberQuantity
ALF053108.22100 g
Catalog number ALF053108.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS23138-50-3
IUPAC Name1-ethyl-4-isocyanatobenzene
Molecular FormulaC9H9NO
InChI KeyFWPYUSLQCQDLJR-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear or slightly turbid colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
CommentMay form slight sediment on storage
Refractive Index1.5250-1.5290 @ 20?C
It is mainly used as an electroplating intermediate. It is also used in medical research.