Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals
Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals
Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals
Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals
CAS: 16924-03-1 | K2NbF7
Have Questions?
Catalog NumberQuantity
ALF051110.1850 g
Catalog number ALF051110.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS17372-87-1
IUPAC Namedisodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6Br4Na2O5
InChI KeySEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10 %
Specific extinction E (1%/1cm)on dried substance
Appearance (Form)Crystalline powder
Specific extinction E (1%/1cm)>=1100 (lambda max, 0.001M Na2CO3)