Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
CAS: 18868-43-4 | MoO2 | 127.948 g/mol
Have Questions?
Catalog NumberQuantity
ALF048117.22100 g
Catalog number ALF048117.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS60-11-7
IUPAC NameN,N-dimethyl-4-(2-phenyldiazen-1-yl)aniline
Molecular FormulaC14H15N3
InChI KeyJCYPECIVGRXBMO-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown
Loss on drying=<3 %
Appearance (Form)Powder
Visual transition intervalfrom pH 2.9 (red) to pH 4.0 (yellow)
Infrared spectrumConforms
View more
Molybdenum(IV) oxide is used to catalyze the dehydrogenation of alcohols. It is also used as an intermediates and processing aids. It finds application in the manufacture of molybdenum nano-wires.