Potassium molybdenum oxide, 95%, Thermo Scientific Chemicals
Potassium molybdenum oxide, 95%, Thermo Scientific Chemicals
Potassium molybdenum oxide, 95%, Thermo Scientific Chemicals
Potassium molybdenum oxide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium molybdenum oxide, 95%, Thermo Scientific Chemicals
CAS: 13446-49-6 | K2MoO4 | 238.14 g/mol
Have Questions?
Catalog NumberQuantity
ALF048112.22100 g
Catalog number ALF048112.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS64248-56-2
IUPAC Name2-bromo-1,3-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyHRZTZLCMURHWFY-UHFFFAOYSA-N
SMILESFC1=CC=CC(F)=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow to pale brown
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5090-1.5130 @ 20°C
Used as an organic intermediate.