Benzyltrimethylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
CAS: 100-85-6 | C10H17NO | 167.252 g/mol
Have Questions?
Catalog NumberQuantity
ALF047189.AP500 mL
Catalog number ALF047189.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzyltrimethylammonium hydroxide
CAS100-85-6
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.