Yttrium, Oil based standard solution, Specpure™ Y 1000μg/g
Product Image
Thermo Scientific Chemicals
Yttrium, Oil based standard solution, Specpure™ Y 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF047172.1850 g
Catalog number ALF047172.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS115-95-7
IUPAC Name3,7-dimethylocta-1,6-dien-3-yl acetate
Molecular FormulaC12H20O2
InChI KeyUWKAYLJWKGQEPM-UHFFFAOYNA-N
SMILESCC(C)=CCCC(C)(OC(C)=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Density(25°C) 0.895 to 0.914 g/cm³
GC>=94 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid