Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Solvent for LC-MS analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
Catalog number ALF047138.M1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 x 1 L
Request bulk or custom format
Chemical Identifiers
CAS5419-55-6
IUPAC Nametris(propan-2-yl) borate
Molecular FormulaC9H21BO3
InChI KeyNHDIQVFFNDKAQU-UHFFFAOYSA-N
SMILESCC(C)OB(OC(C)C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.3755 to 1.3775 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
Solvent for LC-MS analysisAcetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.