Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, LC-MS Grade, 99.8%, Thermo Scientific Chemicals
Solvent for LC-MS analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
Catalog number ALF047138.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS924-44-7
IUPAC Nameethyl 2-oxoacetate
Molecular FormulaC4H6O3
InChI KeyDBPFRRFGLYGEJI-UHFFFAOYSA-N
SMILESCCOC(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolution
CommentExists partly in the polymerised form; depolymerises on heating
Appearance (Color)Clear colorless to pale yellow
Acid-base back titration≥45.0% to ≤55.0% (Aldehyde function)
Solvent for LC-MS analysisAcetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.