n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Catalog NumberQuantity
ALF047104.K21 L
Catalog number ALF047104.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS149022-20-8
IUPAC Namesodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
Molecular FormulaC10H13N5NaO7P
InChI KeyFYWLYWMEGHCZAX-GWKNMROSNA-M
SMILES[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific optical rotation-41° to -43° (20°C, 589 nm) (c=2 in water)
Infrared spectrumConforms
Water=<20.00 %
Appearance (Color)White
View more
n-Hexane is used as a solvent, paint thinner, and chemical reaction medium. It is used to extract edible oils from seeds and vegetables, as a special-use solvent, and as a cleaning agent. Hexanes are commonly used in chromatography as a non-polar solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only