Potassium hydroxide, 45% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 45% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 45% w/v aq. soln., Thermo Scientific Chemicals
Potassium hydroxide, 45% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 45% w/v aq. soln., Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF047010.K21 L
Catalog number ALF047010.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hydroxide
ColorColorless
CAS1310-58-3
Health Hazard 1H290-H302-H314-H335
Health Hazard 2GHS H Statement
H314-H302