Sepabeads∣r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms
Sepabeads∣r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms
Sepabeads∣r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms
Sepabeads∣r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms
Thermo Scientific Chemicals
Sepabeads∣r SP207, synthetic adsorbent resin, highly porous type, brominated PS-DVB, P.R. 110 angstroms
CAS: 87924-01-4 | C18H18 | 234.342 g/mol
Have Questions?
Catalog NumberQuantity
ALF046848.A11 kg
Catalog number ALF046848.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSepabeads™ SP207, synthetic adsorbent resin
Physical Form250μm Beads,S.A. 600m
Name NoteHighly porous type, 120g/L on brominated PS-DVB
CAS87924-01-4
Recommended StorageAmbient temperatures
View more
Seabees SP207, is used in Brominating of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion™ HP20 resin. Sepabeads™ SP-207 was used in kinetic and thermodynamics investigation for adsorption/desorption behavior of anthocyanin pigments. It was used as column packing material for purification of phenolic fraction. It is widely used as a adsorbent for purification in biopharmaceutical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Bromination of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion™ HP20 resin
RUO – Research Use Only