Diaion™ CRB05(OH), chelating resin, highly porous type, 0.95 meq/ml on poly(styrene-divinylbenzene)
Diaion™ CRB05(OH), chelating resin, highly porous type, 0.95 meq/ml on poly(styrene-divinylbenzene)
Thermo Scientific Chemicals
Diaion™ CRB05(OH), chelating resin, highly porous type, 0.95 meq/ml on poly(styrene-divinylbenzene)
CAS: 71799-50-3
Have Questions?
Catalog NumberQuantity
ALF046721.A11 kg
Catalog number ALF046721.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialDiaion™ CRB05 (OH), chelating resin
Physical Form300-850μm Beads
Name NoteHighly porous type, 0.95meq/mL on poly(styrene-divinylbenzene)
CAS71799-50-3
Recommended StorageAmbient temperatures
View more
Boron, arsenic, gallium, and selenium removal