Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
CAS: 124-41-4 | CH3NaO | 54.024 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALF046585.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS134-81-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (GC)≥97.5%
Melting Point (clear melt)92-98?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Sodium methoxide 2% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.