Diaion™ WA21J, ion exchange resin, weakly basic porous type, 2.0 meq/ml on poly(styrene-divinylbenzene)
Diaion™ WA21J, ion exchange resin, weakly basic porous type, 2.0 meq/ml on poly(styrene-divinylbenzene)
Diaion™ WA21J, ion exchange resin, weakly basic porous type, 2.0 meq/ml on poly(styrene-divinylbenzene)
Diaion™ WA21J, ion exchange resin, weakly basic porous type, 2.0 meq/ml on poly(styrene-divinylbenzene)
Thermo Scientific Chemicals
Diaion™ WA21J, ion exchange resin, weakly basic porous type, 2.0 meq/ml on poly(styrene-divinylbenzene)
CAS: 65945-33-7 | C23H31N3 | 349.52 g/mol
Have Questions?
Catalog NumberQuantity
ALF046418.A11 kg
Catalog number ALF046418.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS62476-56-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)117.0-126.0?C
Appearance (Color)White to yellow to brown
Deacidification, solvent treatment, refining of gelatin, adsorption of aldehydes