6-Maleimidohexanoic acid, Thermo Scientific Chemicals
6-Maleimidohexanoic acid, Thermo Scientific Chemicals
6-Maleimidohexanoic acid, Thermo Scientific Chemicals
6-Maleimidohexanoic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
6-Maleimidohexanoic acid, Thermo Scientific Chemicals
CAS: 55750-53-3 | C10H13NO4 | 211.217 g/mol
Have Questions?
Catalog NumberQuantity
ALF046384.065 g
Catalog number ALF046384.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2481-94-9
IUPAC NameN,N-diethyl-4-(2-phenyldiazen-1-yl)aniline
Molecular FormulaC16H19N3
InChI KeySJJISKLXUJVZOA-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Melting Point (clear melt)91.0-98.0°C
FormPowder
Water Content (Karl Fischer Titration)≤2.0%
Assay (GC)≥97.5%
6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.