Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
CAS: 1907-33-1 | C4H9LiO | 80.055 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ALF046366.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS15448-47-2
IUPAC Name(2R)-2-methyloxirane
Molecular FormulaC3H6O
InChI KeyGOOHAUXETOMSMM-GSVOUGTGSA-N
SMILESC[C@@H]1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation13.0 ± 2.0? (neat)
Refractive Index1.3655-1.3685 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
View more
Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.