Tetra-n-butylammonium hydroxide, 1.0M aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 1.0M aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 1.0M aq. soln., Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 1.0M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 1.0M aq. soln., Thermo Scientific Chemicals
CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Catalog NumberdimQuantity
ALF046315.AE
Catalog number ALF046315.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight259.48
Unit Size
Tetra-n-butylammonium hydroxide is used in biological studies for the detection of counter-anion effect on antimicrobial activity of tetrabutylammonium salts, intermediates, Plating agents and surface treating agents, laboratory chemicals.