Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zeolite ZSM-5, ammonium, Thermo Scientific Chemicals
CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF045880.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS24363-37-9
IUPAC Namebarium(2+) bis(propan-2-olate)
Molecular FormulaC6H14BaO2
InChI KeyCPUJSIVIXCTVEI-UHFFFAOYSA-N
SMILES[Ba++].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormLiquid.
Assay (unspecified)(Gravimetric/Titration) 20% active alkoxide
Elemental analysis by ICPTotal trace elements <5000ppm
Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
RUO – Research Use Only