Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
Thermo Scientific Chemicals
Gold nanopowder, APS 20-25nm, 99.95+% (metals basis)
CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Catalog NumberQuantity
ALF045822.031 g
Catalog number ALF045822.03
Price (MYR)
7,280.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
7,280.00
Add to cart
Specifications
Chemical Name or MaterialGold nanopowder
ColorGolden
CAS7440-57-5
Recommended StorageAmbient temperatures
RTECS NumberMD5070000
View more
Gold nanopowder is intercalated within a polymer matrix lower the effective conjugation length of the polymer, resulting in a blue shift in the adsorption. It also finds application in plasmonics, electronics and optical applications.