Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
CAS: 68-04-2 | C6H5Na3O7 | 258.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF045556.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS5423-07-4
IUPAC Namedisodium 3-[(1E)-2-(2,4-dinitrophenyl)diazen-1-yl]-4-hydroxynaphthalene-2,7-disulfonate
Molecular FormulaC16H8N4Na2O11S2
InChI KeyXBLIFEQTVVSTIM-QIKYXUGXSA-L
SMILES[Na+].[Na+].OC1=C2C=CC(=CC2=CC(=C1N=NC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV spectrophotometry>=85 %
Visual transition intervalfrom pH 6.0(bright yellow) to pH 7.2(bright blue)
Appearance (Color)Yellow to orange to brown
Infrared spectrumConforms
Trisodium citrate is used in food additives, beverages, instant drinks, pharmaceuticals and detergents. It is also used as buffering agent, acidity regulator, sequestering or emulsifying agent. It is an effective agent for removal of carbonate scale from boilers and for cleaning automobile radiators. In the medical field, it is used as an osmotic laxative and an active component of the WHO oral rehydration solution. It is also employed to control acidity in some substances such as gelatin desserts. Further, it is used as a flavor or as a preservative. In addition to this, it is used for the preservation of blood in blood banks.