Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trisodium citrate, anhydrous, 99%, Thermo Scientific Chemicals
CAS: 68-04-2 | C6H5Na3O7 | 258.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ALF045556.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTrisodium citrate
Name NoteAnhydrous
CAS68-04-2
Melting Point300°C
ColorWhite
View more
Trisodium citrate is used in food additives, beverages, instant drinks, pharmaceuticals and detergents. It is also used as buffering agent, acidity regulator, sequestering or emulsifying agent. It is an effective agent for removal of carbonate scale from boilers and for cleaning automobile radiators. In the medical field, it is used as an osmotic laxative and an active component of the WHO oral rehydration solution. It is also employed to control acidity in some substances such as gelatin desserts. Further, it is used as a flavor or as a preservative. In addition to this, it is used for the preservation of blood in blood banks.