Devarda's Alloy, granular
Devarda's Alloy, granular
Devarda's Alloy, granular
Devarda's Alloy, granular
Thermo Scientific Chemicals
Devarda's Alloy, granular
CAS: 8049-11-4 | Al9Cu10Zn | 943.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF045526.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS4651-81-4
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0%
Appearance (Color)White to pale brown
FormSolid
Devarda's Alloy is used as reducing agent in analytical chemistry for the determination of nitrates after their reduction to ammonia under alkaline conditions. It is also used in the quantitative or qualitative analysis of nitrates in agriculture and soil science before the development of ion chromatography. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.