Copper nanopowder, APS 20-50nm, 99.9% (metals basis), Thermo Scientific Chemicals
Copper nanopowder, APS 20-50nm, 99.9% (metals basis), Thermo Scientific Chemicals
Copper nanopowder, APS 20-50nm, 99.9% (metals basis), Thermo Scientific Chemicals
Copper nanopowder, APS 20-50nm, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper nanopowder, APS 20-50nm, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF045504.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS64-86-8
IUPAC NameN-[(10S)-3,4,5,14-tetramethoxy-13-oxotricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
Molecular FormulaC22H25NO6
InChI KeyIAKHMKGGTNLKSZ-INIZCTEOSA-N
SMILESCOC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Assay (HPLC)≥94.0% (dry wt. basis)
Loss on Drying≤3.5% (105°C/constant weight)
Appearance (Color)White to pale cream to pale yellow