Silver(I) behenate, Thermo Scientific Chemicals
Silver(I) behenate, Thermo Scientific Chemicals
Silver(I) behenate, Thermo Scientific Chemicals
Silver(I) behenate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver(I) behenate, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF045494.1425 g
Catalog number ALF045494.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver(I) behenate
Melting Point(decomposition)
CAS2489-05-6
ColorWhite
Recommended StorageAmbient temperatures
View more
Silver(I) behenate is used as a calibration standard in X-ray spectrophotometer to an absolute X-ray energy scale. It is utilized for low-angle diffraction standard in X-ray crystallography.