Platinum, 5% on activated carbon paste, B103032-5, Thermo Scientific Chemicals
Platinum, 5% on activated carbon paste, B103032-5, Thermo Scientific Chemicals
Platinum, 5% on activated carbon paste, B103032-5, Thermo Scientific Chemicals
Platinum, 5% on activated carbon paste, B103032-5, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum, 5% on activated carbon paste, B103032-5, Thermo Scientific Chemicals
Material For Gas Sensors
Have Questions?
Catalog NumberQuantity
ALF045445.065 g
Catalog number ALF045445.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.998% (metals basis)
Appearance (Color)Gray
FormFoil, 0.5mm (0.02in) thick.
Platinum, 5% on activated carbon paste is used for a broad range of reactions common to palladium on carbon catalysts. it is utilized for hydrogenolysis reactions using molecular hydrogen.