Arsenic, plasma standard solution, Specpure™ As 10μg/mL
Arsenic, plasma standard solution, Specpure™ As 10μg/mL
Thermo Scientific Chemicals
Arsenic, plasma standard solution, Specpure™ As 10μg/mL
Have Questions?
Catalog NumberQuantity
ALF045246.AE100 mL
Catalog number ALF045246.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS7758-01-2
IUPAC Namepotassium bromate
Molecular FormulaBrKO3
InChI KeyOCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES[K+].[O-][Br](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<50 ppm
Sodium (Na)=<100 ppm
Appearance (Color)White
Loss on drying=<0.1 % (105°C)
pH5 to 9 (5 % at 20°C)