Polypropylene sheet, 3.18mm (0.125in) thick
Polypropylene sheet, 3.18mm (0.125in) thick
Polypropylene sheet, 3.18mm (0.125in) thick
Polypropylene sheet, 3.18mm (0.125in) thick
Thermo Scientific Chemicals
Polypropylene sheet, 3.18mm (0.125in) thick
CAS: 9003-07-0 | (C3H6)n | 354.58 g/mol
Have Questions?
Catalog NumberQuantity
ALF045196.HB300 x 300 mm
Catalog number ALF045196.HB
Price (MYR)
286.00
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (MYR)
286.00
Add to cart
Specifications
Chemical Name or MaterialPolypropylene sheet
Name Note3.18mm (0.125mm) thick
CAS9003-07-0
Recommended StorageAmbient temperatures
TSCAYes