Polypropylene sheet, 6.36mm (0.25in) thick
Polypropylene sheet, 6.36mm (0.25in) thick
Polypropylene sheet, 6.36mm (0.25in) thick
Polypropylene sheet, 6.36mm (0.25in) thick
Thermo Scientific Chemicals
Polypropylene sheet, 6.36mm (0.25in) thick
CAS: 9003-07-0 | (C3H6)n | 354.58 g/mol
Have Questions?
Catalog NumberQuantity
ALF045174.HB300 x 300 mm
Catalog number ALF045174.HB
Price (MYR)
331.00
Add to cart
Quantity:
300 x 300 mm
Request bulk or custom format
Price (MYR)
331.00
Add to cart
Specifications
Chemical Name or MaterialPolypropylene sheet
Name Note6.36mm (0.25 in.) thick
CAS9003-07-0
Recommended StorageAmbient temperatures
RTECS NumberUD1842000