Indium powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Indium powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Indium powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Indium powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Indium powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF045122.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7439-97-6
IUPAC Namemercury
Molecular FormulaHg
InChI KeyQSHDDOUJBYECFT-UHFFFAOYSA-N
SMILES[Hg]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white
Insoluble matter=<0.002 % (in HNO3)
Assay>=99.990 %
Copper (Cu)=<0.1 ppm
Cadmium (Cd)=<0.1 ppm