Silver powder, -120+325 mesh, atomized, 99% (metals basis), Thermo Scientific Chemicals
Silver powder, -120+325 mesh, atomized, 99% (metals basis), Thermo Scientific Chemicals
Silver powder, -120+325 mesh, atomized, 99% (metals basis), Thermo Scientific Chemicals
Silver powder, -120+325 mesh, atomized, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver powder, -120+325 mesh, atomized, 99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF045097.1425 g
Catalog number ALF045097.14
Price (MYR)
1,184.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,184.00
Add to cart
Specifications
Chemical Name or MaterialSilver powder
CAS7440-22-4
Recommended StorageAmbient temperatures
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECS Number231-131-3