Platinum wire, 0.25mm (0.010in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.010in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.010in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.010in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.010in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Platinum wire, 0.25mm (0.010in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Form: Approx.1.05g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 cm
25 cm
50 cm
Catalog number ALF045093.BY
Price (MYR)
2,055.00
Add to cart
Quantity:
100 cm
Request bulk or custom format
Price (MYR)
2,055.00
Add to cart
Specifications
Chemical Name or MaterialPlatinum
ColorWhite
CAS7440-06-4
AppearanceSilvery white wire