Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS, Thermo Scientific Chemicals
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS, Thermo Scientific Chemicals
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS, Thermo Scientific Chemicals
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS, Thermo Scientific Chemicals
CAS: 75-59-2 | C4H13NO | 91.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number ALF044938.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylammonium hydroxide
CAS75-59-2
Health Hazard 1H302-H311-H314-H370-H372
Health Hazard 2GHS H Statement
H301-H311-H314-H371-H373
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
Tetramethylammonium hydroxide is used as anisotropic etching of silicon, as a basic solvent in the development of acidic photo resist in the photolithography process, and is highly effective in stripping photo resist, and is used as a surfactant in the synthesis of ferrofluid, to inhibit nanoparticle aggregation.