Aluminum rod, 6.35mm (0.25in) dia, 99.95% (metals basis)
Aluminum rod, 6.35mm (0.25in) dia, 99.95% (metals basis)
Aluminum rod, 6.35mm (0.25in) dia, 99.95% (metals basis)
Aluminum rod, 6.35mm (0.25in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals
Aluminum rod, 6.35mm (0.25in) dia, 99.95% (metals basis)
CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF044850.G11 m
Catalog number ALF044850.G1
Price (MYR)
503.00
Add to cart
Quantity:
1 m
Request bulk or custom format
Price (MYR)
503.00
Add to cart
Specifications
Chemical Name or MaterialAluminum
CAS7429-90-5
AppearanceSilver-colored rod
Recommended StorageAmbient temperatures
EINECS Number231-072-3