Malonic acid disodium salt, 99%, Thermo Scientific Chemicals
Malonic acid disodium salt, 99%, Thermo Scientific Chemicals
Malonic acid disodium salt, 99%, Thermo Scientific Chemicals
Malonic acid disodium salt, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonic acid disodium salt, 99%, Thermo Scientific Chemicals
CAS: 141-95-7 | C3H2Na2O4 | 148.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF044841.22
Price (MYR)
684.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
684.00
Add to cart
Specifications
Chemical Name or MaterialMalonic acid disodium salt
CAS141-95-7
Recommended StorageAmbient temperatures
RTECS NumberOO1750000
EINECS Number205-514-0
View more
Malonic acid disodium salt is used in organic Synthesis, pharmaceuticals, agrochemicals and dye stuffs.