12-Hydroxystearic acid, 95%, Thermo Scientific Chemicals
12-Hydroxystearic acid, 95%, Thermo Scientific Chemicals
12-Hydroxystearic acid, 95%, Thermo Scientific Chemicals
12-Hydroxystearic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
12-Hydroxystearic acid, 95%, Thermo Scientific Chemicals
CAS: 106-14-9 | C18H36O3 | 300.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALF044810.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS61-76-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Melting point140°C to 145°C
Loss on drying=<1 % (1 g, 105°C)
Infrared spectrumConforms
Appearance (Form)Crystalline powder
View more
12-Hydroxystearic acid is used in cosmetics, wax blends, heavy duty greases, polishes, inks and hot melt adhesives, as a lubricant for natural and synthetic rubbers and as a source for industrial oleo chemicals. It is also used as an intermediate in pharmaceuticals.