Hydroxypropyl methylcellulose, Thermo Scientific Chemicals
Hydroxypropyl methylcellulose, Thermo Scientific Chemicals
Hydroxypropyl methylcellulose, Thermo Scientific Chemicals
Hydroxypropyl methylcellulose, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydroxypropyl methylcellulose, Thermo Scientific Chemicals
CAS: 9004-65-3 | C56H108O30 | 1261.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 kg
Catalog number ALF044779.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHydroxypropyl methylcellulose
CAS9004-65-3
Recommended StorageAmbient temperatures
TSCAYes
Molecular FormulaC56H108O30
View more
Hydroxypropyl methylcellulose is used as an ophthalmic lubricant, an emulsifier and a thickening and suspending agent. It is widely used as an excipient in pharmaceutical formulations. It acts as a food additive. Its eye drops are known as artificial tears, which are used to relieve eye dryness and soreness. It finds applications in various fields as tile adhesives, gypsum products, cosmetics, cement renders, contact lenses, detergents and cleaners.