Palladium nitrate, Matrix Modifier Solution, Specpure™
Palladium nitrate, Matrix Modifier Solution, Specpure™
Thermo Scientific Chemicals
Palladium nitrate, Matrix Modifier Solution, Specpure™
Have Questions?
Catalog NumberQuantity
ALF044765.AE100 mL
Catalog number ALF044765.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS725-89-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)140.0-147.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%