Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Used in phosphatase determinations
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF044711.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS3321-64-0
IUPAC Namebutylphosphonic acid
Molecular FormulaC4H11O3P
InChI KeyUOKRBSXOBUKDGE-UHFFFAOYSA-N
SMILESCCCCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder or flakes
Assay (Aqueous acid-base Titration)>97.5 to <102.5%
Water Content (Karl Fischer Titration)<2.0%
Phenyl phosphate disodium salt is used in phosphatase determinations. It is also used as reagent in chemical research.