Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl phosphate disodium salt hydrate, 98%, Thermo Scientific Chemicals
Used in phosphatase determinations
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF044711.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1308-06-1
IUPAC Namedicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular FormulaCo3O4
InChI KeyUBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Phenyl phosphate disodium salt is used in phosphatase determinations. It is also used as reagent in chemical research.