Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol, Thermo Scientific Chemicals
Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol, Thermo Scientific Chemicals
Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol, Thermo Scientific Chemicals
Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol, Thermo Scientific Chemicals
CAS: 38218-24-5 | C9H21InO3 | 292.082 g/mol
Have Questions?
Catalog NumberQuantity
ALF044562.AC25 mL
Catalog number ALF044562.AC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialIndium (III) isopropoxide
CAS38218-24-5
Health Hazard 1H225-H319-H335-H336
Health Hazard 2GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
View more
Precursor used in the preparation of indium oxide containing films